Product Details

SNP ID
rs7259256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36687445 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGCCACGCACTATGAGAATTTGG[A/G]AGGGGAACCAGAGATCAAAGTTCGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01534 PubMed Links

Gene Details

Gene
LINC01534
Gene Name
long intergenic non-protein coding RNA 1534
There are no transcripts associated with this gene.

Gene
ZNF567
Gene Name
zinc finger protein 567
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300979.1 945 Intron NP_001287908.1
NM_001322911.1 945 Intron NP_001309840.1
NM_001322912.1 945 Intron NP_001309841.1
NM_001322913.1 945 Intron NP_001309842.1
NM_001322914.1 945 Intron NP_001309843.1
NM_001322915.1 945 Intron NP_001309844.1
NM_001322916.1 945 Intron NP_001309845.1
NM_001322917.1 945 Intron NP_001309846.1
NM_001322918.1 945 Intron NP_001309847.1
NM_001322919.1 945 Intron NP_001309848.1
NM_001322920.1 945 Intron NP_001309849.1
NM_152603.4 945 Intron NP_689816.2
XM_011526584.2 945 Intron XP_011524886.1
XM_011526585.2 945 Intron XP_011524887.1
XM_017026416.1 945 Intron XP_016881905.1
XM_017026417.1 945 UTR 5 XP_016881906.1
XM_017026418.1 945 Intron XP_016881907.1
XM_017026419.1 945 Intron XP_016881908.1
XM_017026420.1 945 UTR 5 XP_016881909.1
XM_017026421.1 945 Intron XP_016881910.1

View Full Product Details