Product Details

SNP ID
rs8110668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50421104 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATCCCTTCTTCTGTTGATGACAC[C/T]TGGGCTGTTTCCAGTTGGAGACTCA
Phenotype
MIM: 174761 MIM: 606802
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLD1 PubMed Links

Gene Details

Gene
POLD1
Gene Name
DAN polymerase delta 1, catalytic subunit
There are no transcripts associated with this gene.

Gene
SPIB
Gene Name
Spi-B transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243998.1 Intron NP_001230927.1
NM_001243999.1 Intron NP_001230928.1
NM_001244000.1 Intron NP_001230929.1
NM_003121.4 Intron NP_003112.2

View Full Product Details