Product Details

SNP ID
rs10445585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52379571 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCGCGCCTGGCCTCATTTTTAA[A/T]TTTTTTAGAGACAGGGTTTCACTCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF528-AS1 PubMed Links

Gene Details

Gene
ZNF528-AS1
Gene Name
ZNF528 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZNF880
Gene Name
zinc finger protein 880
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145434.1 Intron NP_001138906.1
XM_017026812.1 Intron XP_016882301.1

View Full Product Details