Product Details

SNP ID
rs13000658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202230180 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGATACATACTGTATGATTTCAC[C/T]TATAAAATGTTATGCTGTATAAAAA
Phenotype
MIM: 601912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUMO1 PubMed Links

Gene Details

Gene
SUMO1
Gene Name
small ubiquitin-like modifier 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005781.1 Intron NP_001005781.1
NM_001005782.1 Intron NP_001005782.1
NM_003352.4 Intron NP_003343.1

View Full Product Details