Product Details

SNP ID
rs7247121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21978688 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCCTCCAAAGGATCACAACTCAT[C/T]GCCAGCAAAGAAAGAAAACTGAATG
Phenotype
MIM: 603977
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF208 PubMed Links

Gene Details

Gene
ZNF208
Gene Name
zinc finger protein 208
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007153.3 Intron NP_009084.2
XM_017027257.1 Intron XP_016882746.1

View Full Product Details