Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020747.2 | Intron | NP_065798.2 | |||
XM_005272037.2 | Intron | XP_005272094.1 | |||
XM_005272038.1 | Intron | XP_005272095.1 | |||
XM_005272039.2 | Intron | XP_005272096.1 | |||
XM_005272040.3 | Intron | XP_005272097.1 | |||
XM_011543520.2 | Intron | XP_011541822.1 | |||
XM_011543522.2 | Intron | XP_011541824.1 | |||
XM_011543523.2 | Intron | XP_011541825.1 | |||
XM_017009674.1 | Intron | XP_016865163.1 |