Product Details

SNP ID
rs4973768
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:27374522 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCACTTAAAACAAGCAGTTAATTAC[C/T]TAAACATGAGTTACCTTTGCTCTTA
Phenotype
MIM: 603353
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEK10 PubMed Links

Gene Details

Gene
NEK10
Gene Name
NIMA related kinase 10
There are no transcripts associated with this gene.

Gene
SLC4A7
Gene Name
solute carrier family 4 member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258379.1 5586 UTR 3 NP_001245308.1
NM_001258380.1 5586 UTR 3 NP_001245309.1
NM_001321103.1 5586 UTR 3 NP_001308032.1
NM_001321104.1 5586 UTR 3 NP_001308033.1
NM_001321105.1 5586 UTR 3 NP_001308034.1
NM_001321106.1 5586 UTR 3 NP_001308035.1
NM_001321107.1 5586 UTR 3 NP_001308036.1
NM_001321108.1 5586 UTR 3 NP_001308037.1
NM_003615.4 5586 UTR 3 NP_003606.3
XM_005265598.4 5586 UTR 3 XP_005265655.1
XM_005265600.4 5586 Intron XP_005265657.1
XM_006713421.2 5586 Intron XP_006713484.1
XM_011534256.2 5586 Intron XP_011532558.1
XM_011534258.1 5586 UTR 3 XP_011532560.1
XM_011534261.2 5586 UTR 3 XP_011532563.1
XM_011534263.2 5586 Intron XP_011532565.1
XM_011534265.1 5586 UTR 3 XP_011532567.1
XM_017007526.1 5586 Intron XP_016863015.1
XM_017007527.1 5586 Intron XP_016863016.1
XM_017007528.1 5586 UTR 3 XP_016863017.1
XM_017007529.1 5586 UTR 3 XP_016863018.1

View Full Product Details