Product Details

SNP ID
rs11981330
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:150382371 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTATTTCCGTTCACCTGCGCCTTCA[A/G]GAACCTGAGCTCAAAGCGCTGCAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
REPIN1 PubMed Links

Gene Details

Gene
REPIN1
Gene Name
replication initiator 1
There are no transcripts associated with this gene.

Gene
ZNF775
Gene Name
zinc finger protein 775
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173680.3 Intron NP_775951.2

View Full Product Details