Product Details

SNP ID
rs6948729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100189749 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGACCCATCCTGTAAGGGGTCATT[A/G]CATGCTCATCTTGTTTCTGGGTTGT
Phenotype
MIM: 608489
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GATS PubMed Links

Gene Details

Gene
GATS
Gene Name
GATS, stromal antigen 3 opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178831.6 Intron NP_849153.3
XM_017012153.1 Intron XP_016867642.1
XM_017012154.1 Intron XP_016867643.1
XM_017012155.1 Intron XP_016867644.1
XM_017012156.1 Intron XP_016867645.1
XM_017012157.1 Intron XP_016867646.1
XM_017012158.1 Intron XP_016867647.1
XM_017012159.1 Intron XP_016867648.1
Gene
STAG3
Gene Name
stromal antigen 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282716.1 Intron NP_001269645.1
NM_001282717.1 Intron NP_001269646.1
NM_001282718.1 Intron NP_001269647.1
NM_012447.3 Intron NP_036579.2
XM_011515742.1 Intron XP_011514044.1
XM_017011683.1 Intron XP_016867172.1
XM_017011684.1 Intron XP_016867173.1
XM_017011685.1 Intron XP_016867174.1
XM_017011686.1 Intron XP_016867175.1
XM_017011687.1 Intron XP_016867176.1

View Full Product Details