Product Details

SNP ID
rs4653718
Assay Type
Validated
NCBI dbSNP Submissions
47
Location
Chr.1:226148462 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATACCTATAATTAAGCATGTTTCAC[G/A]TGCAAATGAGAAATACAGAACCATC
Phenotype
MIM: 606809
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ACBD3 PubMed Links

Gene Details

Gene
ACBD3
Gene Name
acyl-CoA binding domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022735.3 Intron NP_073572.2

View Full Product Details