Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001305077.1 | 1016 | UTR 3 | NP_001292006.1 | ||
NM_001305078.1 | 1016 | UTR 3 | NP_001292007.1 | ||
NM_001305079.1 | 1016 | UTR 3 | NP_001292008.1 | ||
NM_001305080.1 | 1016 | UTR 3 | NP_001292009.1 | ||
NM_001305081.1 | 1016 | UTR 3 | NP_001292010.1 | ||
NM_001305082.1 | 1016 | UTR 3 | NP_001292011.1 | ||
NM_001305083.1 | 1016 | UTR 3 | NP_001292012.1 | ||
NM_001305084.1 | 1016 | UTR 3 | NP_001292013.1 | ||
NM_001305085.1 | 1016 | UTR 3 | NP_001292014.1 | ||
NM_001305086.1 | 1016 | UTR 3 | NP_001292015.1 | ||
NM_001305087.1 | 1016 | UTR 3 | NP_001292016.1 | ||
NM_001305088.1 | 1016 | UTR 3 | NP_001292017.1 | ||
NM_015654.4 | 1016 | UTR 3 | NP_056469.2 | ||
XM_005257212.4 | 1016 | UTR 3 | XP_005257269.1 | ||
XM_006721813.3 | 1016 | UTR 3 | XP_006721876.1 | ||
XM_011524607.2 | 1016 | Intron | XP_011522909.1 | ||
XM_017024437.1 | 1016 | UTR 3 | XP_016879926.1 | ||
XM_017024438.1 | 1016 | Intron | XP_016879927.1 | ||
XM_017024439.1 | 1016 | UTR 3 | XP_016879928.1 | ||
XM_017024440.1 | 1016 | Intron | XP_016879929.1 | ||
XM_017024441.1 | 1016 | UTR 3 | XP_016879930.1 |