Product Details

SNP ID
rs9900142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:62140530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGTCTTGGTCTTGCTCCTGGGGG[C/T]TGGCGGATGAGCTCCTCCTGGGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLRMTP1 PubMed Links

Gene Details

Gene
POLRMTP1
Gene Name
RNA polymerase mitochondrial pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details