Product Details
- SNP ID
-
rs2040906
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149841061 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGAAAGGTGCTCACCCTTCCCAAA[G/T]ATAGTTGTTTTCTAATATGGGAAAT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SSPO
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79886296] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SSPO
- Gene Name
- SCO-spondin
There are no transcripts associated with this gene.
- Gene
- ZNF862
- Gene Name
- zinc finger protein 862
View Full Product Details