Product Details

SNP ID
rs1357972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123459169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAAACCCTACATAGATACGGTA[C/T]TGAAATTAAATCACATCTAATGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IQUB PubMed Links

Gene Details

Gene
IQUB
Gene Name
IQ motif and ubiquitin domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282855.1 Intron NP_001269784.1
NM_001321293.1 Intron NP_001308222.1
NM_178827.4 Intron NP_849149.3
XM_005250161.3 Intron XP_005250218.1
XM_005250162.4 Intron XP_005250219.1
XM_011515833.2 Intron XP_011514135.1
XM_011515834.2 Intron XP_011514136.1
XM_017011771.1 Intron XP_016867260.1
XM_017011772.1 Intron XP_016867261.1

View Full Product Details