Product Details

SNP ID
rs1855875
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.10:47632721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTTATTTACCATTTAATGTATT[C/T]TAATGTACGTTTTTGCCATATTTAA
Phenotype
MIM: 602396
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ANXA8 PubMed Links

Gene Details

Gene
ANXA8
Gene Name
annexin A8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040084.2 Intron NP_001035173.1
NM_001271702.1 Intron NP_001258631.1
NM_001271703.1 Intron NP_001258632.1
XM_006717951.3 Intron XP_006718014.1
XM_011540096.1 Intron XP_011538398.1
XM_011540098.1 Intron XP_011538400.1
XM_011540099.1 Intron XP_011538401.1
XM_011540101.2 Intron XP_011538403.1

View Full Product Details