Product Details

SNP ID
rs1821336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5488691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAATACTATTCAGAAAGAGCCCT[G/T]CATTTGCCAGGAAGAATGTCAGATT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 7 Intron NP_001005567.2
Gene
OR52D1
Gene Name
olfactory receptor family 52 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005163.2 7 UTR 5 NP_001005163.1

View Full Product Details