Product Details

SNP ID
rs35778288
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:10572872 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTGGGTCCTAATTCAAGGGCCAGGT[A/G]AGCATCCCAGAAACAGAGGCCAGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD33B PubMed Links

Gene Details

Gene
ANKRD33B
Gene Name
ankyrin repeat domain 33B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164440.1 Intron NP_001157912.1
XM_005248343.4 Intron XP_005248400.1
XM_005248344.3 Intron XP_005248401.1

View Full Product Details