Product Details

SNP ID
hCV11656518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45899874 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCAGAATGGTTGGAATAGATTC[A/G]GAACCTGCGGGAAAGAAAGGAGCCC
Phenotype
MIM: 613111 MIM: 608597 MIM: 172425
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTSA PubMed Links

Gene Details

Gene
CTSA
Gene Name
cathepsin A
There are no transcripts associated with this gene.

Gene
NEURL2
Gene Name
neuralized E3 ubiquitin protein ligase 2
There are no transcripts associated with this gene.

Gene
PLTP
Gene Name
phospholipid transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242920.1 1192 Nonsense Mutation CGA,TGA R299* NP_001229849.1
NM_001242921.1 1192 Nonsense Mutation CGA,TGA R306* NP_001229850.1
NM_006227.3 1192 Nonsense Mutation CGA,TGA R394* NP_006218.1
NM_182676.2 1192 Nonsense Mutation CGA,TGA R342* NP_872617.1

View Full Product Details