Product Details

SNP ID
rs7920095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:35014406 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAATTTTTAGAATAACAACTTCT[A/G]TATCAGGCTTAAGAAAGCCACTGAT
Phenotype
MIM: 603135
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUL2 PubMed Links

Gene Details

Gene
CUL2
Gene Name
cullin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198777.1 Intron NP_001185706.1
NM_001198778.1 Intron NP_001185707.1
NM_001198779.1 Intron NP_001185708.1
NM_001324375.1 Intron NP_001311304.1
NM_001324376.1 Intron NP_001311305.1
NM_003591.3 Intron NP_003582.2
XM_011519743.1 Intron XP_011518045.1
XM_011519744.1 Intron XP_011518046.1
XM_011519745.1 Intron XP_011518047.1
XM_011519747.1 Intron XP_011518049.1

View Full Product Details