Product Details

SNP ID
rs5599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:49506301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATTTAAAGGGTCTCAATAAACA[A/G]GACTTTATATTAATAAAAATAGCAT
Phenotype
MIM: 601699
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTGIS PubMed Links

Gene Details

Gene
PTGIS
Gene Name
prostaglandin I2 (prostacyclin) synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000961.3 3176 UTR 3 NP_000952.1

View Full Product Details