Product Details

SNP ID
rs1890985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104139520 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGCCTCGGATGGGAGGGGCCTG[A/G]GCTGGCGTTCCTCCCCCAGCCCAGA
Phenotype
MIM: 613231
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF26A PubMed Links

Gene Details

Gene
KIF26A
Gene Name
kinesin family member 26A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015656.1 Intron NP_056471.1
XM_011536641.2 Intron XP_011534943.1
Gene
LOC105370708
Gene Name
uncharacterized LOC105370708
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017021846.1 Intron XP_016877335.1

View Full Product Details