Product Details

SNP ID
rs1962
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:23045859 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTTTGAAAATCAGAGATGGTGCCA[C/T]CACCAGACAACACAAGTGCTGGGGT
Phenotype
MIM: 182454 MIM: 188040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SSTR4 PubMed Links

Gene Details

Gene
SSTR4
Gene Name
somatostatin receptor 4
There are no transcripts associated with this gene.

Gene
THBD
Gene Name
thrombomodulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000361.2 3806 UTR 3 NP_000352.1

View Full Product Details