Product Details
- SNP ID
-
rs2054166
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:51682713 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTAATTGACAGAAATTAGGACGGGA[A/T]ATCTAGTTTTGGTAAATTCTATGCC
- Phenotype
-
MIM: 611796
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SCG3
PubMed Links
Gene Details
- Gene
- SCG3
- Gene Name
- secretogranin III
View Full Product Details