Product Details

SNP ID
rs230968
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15306759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTCTGCAGAGATTTTTGGAGAA[C/T]GAAACATCAGTTCAGCTTCTCCTAA
Phenotype
MIM: 612683
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEKT3 PubMed Links

Gene Details

Gene
TEKT3
Gene Name
tektin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031898.2 Intron NP_114104.1
XM_011523988.2 Intron XP_011522290.1
XM_011523989.1 Intron XP_011522291.1
XM_011523990.2 Intron XP_011522292.1
XM_017024954.1 Intron XP_016880443.1
XM_017024955.1 Intron XP_016880444.1
XM_017024956.1 Intron XP_016880445.1

View Full Product Details