Product Details
- SNP ID
-
rs1049216
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:184628935 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AGTGGGGGGAATATCATAAAAATTC[G/A]TTACTTCAATGTTTAACTTTTTCAC
- Phenotype
-
MIM: 600636
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CASP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4647694] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CASP3
- Gene Name
- caspase 3
- Gene
- LVCAT8
- Gene Name
- liver cancer-associated transcript 8
There are no transcripts associated with this gene.
View Full Product Details