Product Details

SNP ID
rs1851682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187698301 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTACACCCTTTTCCCCTTCCCCC[A/C]ATTATTGTCACATCCAGTGAAAGAA
Phenotype
MIM: 609138
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC100131635 PubMed Links

Gene Details

Gene
LOC100131635
Gene Name
hCG1645011-like
There are no transcripts associated with this gene.

Gene
RTP2
Gene Name
receptor transporter protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004312.2 1449 UTR 3 NP_001004312.2
XM_017006301.1 1449 UTR 3 XP_016861790.1
XM_017006302.1 1449 UTR 3 XP_016861791.1

View Full Product Details