Product Details

SNP ID
rs1880095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187699866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGAGTCCCAGTCTCAGGAAGTGT[G/T]GTGGGTGGTAGCCTGGGGCTCATCT
Phenotype
MIM: 609138
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100131635 PubMed Links

Gene Details

Gene
LOC100131635
Gene Name
hCG1645011-like
There are no transcripts associated with this gene.

Gene
RTP2
Gene Name
receptor transporter protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004312.2 Intron NP_001004312.2
XM_017006301.1 Intron XP_016861790.1
XM_017006302.1 Intron XP_016861791.1

View Full Product Details