Product Details

SNP ID
rs2305796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12843327 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGACCGTGGGGAGGAGAGTGGAGA[C/T]GGATCCTCCCTCCAATTCCCGGTGC
Phenotype
MIM: 612256 MIM: 609553
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAST1 PubMed Links

Gene Details

Gene
MAST1
Gene Name
microtubule associated serine/threonine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014975.2 Intron NP_055790.1
XM_011527805.2 Intron XP_011526107.1
XM_011527808.2 Intron XP_011526110.1
XM_017026501.1 Intron XP_016881990.1
Gene
MIR6794
Gene Name
microRNA 6794
There are no transcripts associated with this gene.

Gene
RTBDN
Gene Name
retbindin
There are no transcripts associated with this gene.

View Full Product Details