Product Details

SNP ID
rs11575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:131949528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCTCAAACTTGATAGGCTTGGA[A/G]ATTTTGGGAGTACGGATGCACTTTT
Phenotype
MIM: 121009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTGF PubMed Links

Gene Details

Gene
CTGF
Gene Name
connective tissue growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001901.2 992 Silent Mutation ATC,ATT I262I NP_001892.1

View Full Product Details