Product Details

SNP ID
rs2343616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17218540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTGGATGCTCTTCCCAGAAGT[A/C]AAAGGCCAGGCGCAGTGGCTCACGC
Phenotype
MIM: 602129 MIM: 610675
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MYO9B PubMed Links

Gene Details

Gene
MYO9B
Gene Name
myosin IXB
There are no transcripts associated with this gene.

Gene
OCEL1
Gene Name
occludin/ELL domain containing 1
There are no transcripts associated with this gene.

Gene
USE1
Gene Name
unconventional SNARE in the ER 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018467.3 Intron NP_060937.1
XM_017026975.1 Intron XP_016882464.1
XM_017026976.1 Intron XP_016882465.1

View Full Product Details