Product Details
- SNP ID
-
rs7251728
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57289020 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTTCAGATTGTGTCTTTTTCTAGT[A/G]TTTTCAGGTTATTGTTTCTCAGGGT
- Phenotype
-
MIM: 604755
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105372476
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200800476] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105372476
- Gene Name
- uncharacterized LOC105372476
There are no transcripts associated with this gene.
- Gene
- ZNF460
- Gene Name
- zinc finger protein 460
View Full Product Details