Product Details

SNP ID
rs1989866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:111901465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACGTGAAGAACATGCTGTAGACA[G/T]GATGATATTATCATTCATTATACTT
Phenotype
MIM: 606723
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAM1A PubMed Links

Gene Details

Gene
ADAM1A
Gene Name
ADAM metallopeptidase domain 1A (pseudogene)
There are no transcripts associated with this gene.

Gene
MAPKAPK5
Gene Name
mitogen-activated protein kinase-activated protein kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003668.3 8412 Intron NP_003659.2
NM_139078.2 8412 Intron NP_620777.1
XM_017020125.1 8412 Intron XP_016875614.1
XM_017020126.1 8412 Intron XP_016875615.1
XM_017020127.1 8412 UTR 3 XP_016875616.1
XM_017020128.1 8412 UTR 3 XP_016875617.1
XM_017020129.1 8412 Intron XP_016875618.1
XM_017020130.1 8412 Intron XP_016875619.1
XM_017020131.1 8412 Intron XP_016875620.1
XM_017020132.1 8412 UTR 3 XP_016875621.1
XM_017020133.1 8412 UTR 3 XP_016875622.1
XM_017020134.1 8412 UTR 3 XP_016875623.1
XM_017020135.1 8412 UTR 3 XP_016875624.1
XM_017020136.1 8412 UTR 3 XP_016875625.1
XM_017020137.1 8412 Intron XP_016875626.1
XM_017020138.1 8412 UTR 3 XP_016875627.1
XM_017020139.1 8412 Intron XP_016875628.1
Gene
TMEM116
Gene Name
transmembrane protein 116
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193453.1 8412 Intron NP_001180382.1
NM_001193531.1 8412 Intron NP_001180460.1
NM_001294314.1 8412 Intron NP_001281243.1
NM_138341.2 8412 Intron NP_612350.1
XM_006719678.3 8412 Intron XP_006719741.1
XM_011538946.1 8412 Intron XP_011537248.1
XM_011538947.1 8412 Intron XP_011537249.1
XM_011538948.2 8412 Intron XP_011537250.1
XM_011538949.2 8412 Intron XP_011537251.1
XM_011538950.1 8412 Intron XP_011537252.1
XM_011538951.2 8412 Intron XP_011537253.1
XM_011538953.1 8412 Intron XP_011537255.1
XM_011538955.1 8412 Intron XP_011537257.1
XM_011538957.1 8412 Intron XP_011537259.1
XM_011538958.1 8412 Intron XP_011537260.1
XM_017020187.1 8412 Intron XP_016875676.1
XM_017020188.1 8412 Intron XP_016875677.1
XM_017020189.1 8412 Intron XP_016875678.1
XM_017020190.1 8412 Intron XP_016875679.1
XM_017020191.1 8412 Intron XP_016875680.1
XM_017020192.1 8412 Intron XP_016875681.1
XM_017020193.1 8412 Intron XP_016875682.1
XM_017020194.1 8412 Intron XP_016875683.1

View Full Product Details