Product Details

SNP ID
rs11670216
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37012526 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAGAGTGGTGGAGGTCCGACCAG[C/G]ATGATGAAGCCACAGGCGGAGTCTG
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ZNF420 PubMed Links

Gene Details

Gene
ZNF420
Gene Name
zinc finger protein 420
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006723038.3 Intron XP_006723101.1
XM_006723039.3 Intron XP_006723102.1
XM_011526503.2 Intron XP_011524805.1
XM_011526504.2 Intron XP_011524806.1
XM_011526505.2 Intron XP_011524807.1
XM_011526507.2 Intron XP_011524809.1
XM_011526508.2 Intron XP_011524810.1
XM_011526510.2 Intron XP_011524812.1
XM_011526511.2 Intron XP_011524813.1
XM_017026336.1 Intron XP_016881825.1
XM_017026337.1 Intron XP_016881826.1
XM_017026338.1 Intron XP_016881827.1
XM_017026339.1 Intron XP_016881828.1
XM_017026340.1 Intron XP_016881829.1

View Full Product Details