Product Details

SNP ID
rs1888435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:28572778 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATGTTTTTACAAAGTACCAAACA[C/T]TTATTGAACAACTACTATATGCCAG
Phenotype
MIM: 614553
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
N6AMT1 PubMed Links

Gene Details

Gene
N6AMT1
Gene Name
N-6 adenine-specific DNA methyltransferase 1 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013240.5 Intron NP_037372.4
NM_182749.4 Intron NP_877426.4

View Full Product Details