Product Details

SNP ID
rs1926185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88882785 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATCCCTGAAGATATTAGAGCCAG[A/C]TTTCCATTCCTTAGGGCTGATGCAT
Phenotype
MIM: 612352
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
STAMBPL1 PubMed Links

Gene Details

Gene
STAMBPL1
Gene Name
STAM binding protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020799.3 Intron NP_065850.1
XM_006717928.2 Intron XP_006717991.1
XM_006717930.3 Intron XP_006717993.1
XM_011539983.1 Intron XP_011538285.1
XM_011539985.1 Intron XP_011538287.1
XM_017016453.1 Intron XP_016871942.1
XM_017016454.1 Intron XP_016871943.1
XM_017016455.1 Intron XP_016871944.1
XM_017016456.1 Intron XP_016871945.1
XM_017016457.1 Intron XP_016871946.1
XM_017016458.1 Intron XP_016871947.1

View Full Product Details