Product Details

SNP ID
rs9621390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32124265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGGGAGCACTGTGGGACTGGC[C/T]CGCCTGTCCCAGCCTGGCCTGGGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP1B1P1 PubMed Links

Gene Details

Gene
AP1B1P1
Gene Name
adaptor related protein complex 1 beta 1 subunit pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details