Product Details

SNP ID
rs12591859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34085445 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTGACTTTAGTTCATTCATACAT[A/G]AAAAAGATAGCAAATACTATATTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMC7 PubMed Links

Gene Details

Gene
EMC7
Gene Name
ER membrane protein complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020154.2 Intron NP_064539.1

View Full Product Details