Product Details
- SNP ID
-
rs36077761
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:179348666 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AGATTTCACTTATTTTAGTGAAATA[C/T]CATTCAAAAGTCGTCTCTAACTGTC
- Phenotype
-
MIM: 608506
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MFN1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9816602] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MFN1
- Gene Name
- mitofusin 1
View Full Product Details