Product Details
- SNP ID
-
rs13432533
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:26348543 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATTGATTTTTGGTATGGAAGAGC[A/G]TGGGATGGATTCTGTGTTATGTATT
- Phenotype
-
MIM: 607915
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADGRF3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76708830] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADGRF3
- Gene Name
- adhesion G protein-coupled receptor F3
There are no transcripts associated with this gene.
- Gene
- EPT1
- Gene Name
- ethanolaminephosphotransferase 1
There are no transcripts associated with this gene.
View Full Product Details