Product Details

SNP ID
rs35150598
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:136870546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAAATGCTTGATCATTTGCAAG[G/T]ACTACATGCTCTTGAATAATATATT
Phenotype
MIM: 600508
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
NCK1 PubMed Links

Gene Details

Gene
NCK1
Gene Name
NCK adaptor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190796.2 Intron NP_001177725.1
NM_001291999.1 Intron NP_001278928.1
NM_006153.5 Intron NP_006144.1
Gene
NCK1-AS1
Gene Name
NCK1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details