Product Details

SNP ID
rs13283282
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:128703202 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACCCCAACCAGCCCGTGAGCGAGGA[C/G]GGCTGAGTCCGCAGTGTGCGCCACC
Phenotype
MIM: 610714 MIM: 600960
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PKN3 PubMed Links

Gene Details

Gene
PKN3
Gene Name
protein kinase N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317926.1 240 Intron NP_001304855.1
NM_013355.4 240 Intron NP_037487.2
XM_005251946.3 240 UTR 5 XP_005252003.1
XM_006717080.2 240 UTR 5 XP_006717143.1
XM_017014649.1 240 UTR 5 XP_016870138.1
XM_017014650.1 240 Intron XP_016870139.1
Gene
SET
Gene Name
SET nuclear proto-oncogene
There are no transcripts associated with this gene.

View Full Product Details