Product Details

SNP ID
rs3816094
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:121459755 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTCACTCCCAGAACCACATCCTTC[C/T]AGTTGATCCTGCCTCCTACCAATCT
Phenotype
MIM: 104175
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GGTA1P PubMed Links

Gene Details

Gene
GGTA1P
Gene Name
glycoprotein, alpha-galactosyltransferase 1 pseudogene
There are no transcripts associated with this gene.

View Full Product Details