Product Details
- SNP ID
-
rs600952
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:19268871 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTACAATAAAGTGCAGACAGAAGG[A/G]ATTATATAGAACATATATAACTGAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD26P3
PubMed Links
Gene Details
- Gene
- ANKRD26P3
- Gene Name
- ankyrin repeat domain 26 pseudogene 3
There are no transcripts associated with this gene.
View Full Product Details