Product Details

SNP ID
rs5982920
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:2916682 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATATGGGAGAGCTAAAAAAAAAT[C/T]GATCACATGAAGATAGAGAATAGAA
Phenotype
MIM: 300002
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ARSD PubMed Links

Gene Details

Gene
ARSD
Gene Name
arylsulfatase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001669.3 Intron NP_001660.2
NM_009589.3 Intron NP_033667.2
XM_005274514.2 Intron XP_005274571.1
XM_005274515.2 Intron XP_005274572.1

View Full Product Details