Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_003611.2 | Intron | NP_003602.1 | |||
XM_005274604.2 | Intron | XP_005274661.1 | |||
XM_005274606.3 | Intron | XP_005274663.1 | |||
XM_011545592.2 | Intron | XP_011543894.1 | |||
XM_011545594.2 | Intron | XP_011543896.1 | |||
XM_011545595.1 | Intron | XP_011543897.1 | |||
XM_011545597.1 | Intron | XP_011543899.1 | |||
XM_011545598.1 | Intron | XP_011543900.1 | |||
XM_017029909.1 | Intron | XP_016885398.1 | |||
XM_017029910.1 | Intron | XP_016885399.1 | |||
XM_017029911.1 | Intron | XP_016885400.1 |