Product Details

SNP ID
rs11999
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:117820203 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAAGCATGAGGTGGCACGAGGCAGG[A/C]GTTGGCGATGCCACCTGGGGGTCAC
Phenotype
MIM: 601814
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FXYD2 PubMed Links

Gene Details

Gene
FXYD2
Gene Name
FXYD domain containing ion transport regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001680.4 1056 UTR 3 NP_001671.2
NM_021603.3 1056 UTR 3 NP_067614.1
Gene
FXYD6-FXYD2
Gene Name
FXYD6-FXYD2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204268.1 1056 UTR 3 NP_001191197.1
NM_001243598.2 1056 UTR 3 NP_001230527.1

View Full Product Details