Product Details

SNP ID
rs13699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119308942 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAATTCACCTGAACTCGAAAACAG[C/T]GCCCAGCTTCCTCACCGCAGGCACG
Phenotype
MIM: 165360 MIM: 155735
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CBL PubMed Links

Gene Details

Gene
CBL
Gene Name
Cbl proto-oncogene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005188.3 2820 Intron NP_005179.2
XM_011543057.2 2820 Intron XP_011541359.1
XM_017018511.1 2820 Intron XP_016874000.1
Gene
MCAM
Gene Name
melanoma cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006500.2 2820 UTR 3 NP_006491.2
XM_017017759.1 2820 UTR 3 XP_016873248.1
XM_017017760.1 2820 UTR 3 XP_016873249.1
XM_017017761.1 2820 UTR 3 XP_016873250.1
XM_017017762.1 2820 UTR 3 XP_016873251.1
Gene
MIR6756
Gene Name
microRNA 6756
There are no transcripts associated with this gene.

View Full Product Details