Product Details

SNP ID
rs2040620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13721524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCGGGAGTTACAGTCAGTCTCCT[A/G]GAACATACATCTGAGGTGTGTTAGA
Phenotype
MIM: 300202
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC2 PubMed Links

Gene Details

Gene
TRAPPC2
Gene Name
trafficking protein particle complex 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011658.3 Intron NP_001011658.1
NM_001128835.2 Intron NP_001122307.2
NM_014563.5 Intron NP_055378.1
XM_011545565.1 Intron XP_011543867.1
XM_011545566.2 Intron XP_011543868.1

View Full Product Details