Product Details

SNP ID
rs12874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20403940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACGCAAGTGCTGCTGTGCCGCCA[C/G]GCCCAGGGCTATGATCCAAAGTGAC
Phenotype
MIM: 609877
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CRYL1 PubMed Links

Gene Details

Gene
CRYL1
Gene Name
crystallin lambda 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015974.2 1055 UTR 3 NP_057058.2
XM_005266415.3 1055 UTR 3 XP_005266472.1
XM_005266416.4 1055 Intron XP_005266473.1
XM_017020624.1 1055 UTR 3 XP_016876113.1

View Full Product Details