Product Details

SNP ID
rs2438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74447588 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCACGATTTTATACATAGAAAG[C/G]AAACCCATTTACAAAAGAGGCTTGT
Phenotype
MIM: 600753
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLG1 PubMed Links

Gene Details

Gene
GLG1
Gene Name
golgi glycoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145666.1 8097 UTR 3 NP_001139138.1
NM_001145667.1 8097 UTR 3 NP_001139139.1
NM_012201.5 8097 UTR 3 NP_036333.2

View Full Product Details